Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958594.1 | XP_027814395.1 | 10 - 11 | 851 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |