Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNDIG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972017.1 | XP_027827818.1 | 3 - 4 | 743 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |