Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019125.3 | XP_004019174.1 | 2 - 3 | 306 | 72 | |
XM_012101260.2 | XP_011956650.1 | 2 - 3 | 202 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |