Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976826.1 | XP_027832627.1 | 3 - 4 | 974 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |