Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015395.3 | XP_004015444.2 | 5 - 6 | 795 | 103 | |
XM_012094665.3 | XP_011950055.2 | 5 - 6 | 797 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |