Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007963.4 | XP_004008012.2 | 3 - 4 | 1072 | 81 | |
XM_027968679.1 | XP_027824480.1 | 3 - 4 | 1072 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |