Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961047.1 | XP_027816848.1 | 4 - 5 | 600 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |