Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964784.1 | XP_027820585.1 | 4 - 5 | 565 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |