Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184103.2 | XP_012039493.1 | 3 - 4 | 274 | 109 | |
XM_015097914.2 | XP_014953400.1 | 3 - 4 | 779 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |