Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004147.4 | XP_004004196.1 | 2 - 3 | 154 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |