Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970743.1 | XP_027826544.1 | 3 - 4 | 639 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |