Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003220.4 | XP_004003269.2 | 2 - 3 | 1611 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |