Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977588.1 | XP_027833389.1 | 2 - 3 | 409 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |