Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002134.3 | XP_004002183.1 | 3 - 4 | 266 | 64 | |
XM_004002136.4 | XP_004002185.1 | 3 - 4 | 290 | 64 | ||
XM_027971495.1 | XP_027827296.1 | 3 - 4 | 981 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |