Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958479.1 | XP_027814280.1 | 5 - 6 | 627 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |