Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976392.1 | XP_027832193.1 | 4 - 5 | 842 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |