Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004438.4 | XP_004004487.2 | 3 - 4 | 692 | 60 | |
XM_027964602.1 | XP_027820403.1 | 3 - 4 | 693 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |