Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012107098.2 | XP_011962488.1 | 5 - 6 | 822 | 93 | |
XM_012107100.2 | XP_011962490.1 | 4 - 5 | 624 | 93 | ||
XM_027962984.1 | XP_027818785.1 | 4 - 5 | 779 | 93 | ||
XM_027962988.1 | XP_027818789.1 | 5 - 6 | 1433 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |