Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977938.1 | XP_027833739.1 | 2 - 3 | 469 | 72 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Ovis aries |