Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184387.2 | XP_012039777.2 | 4 - 5 | 1153 | 112 | |
XM_027973535.1 | XP_027829336.1 | 4 - 5 | 1030 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |