Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003266.4 | XP_004003315.1 | 5 - 6 | 866 | 125 | |
XM_012101604.3 | XP_011956994.1 | 5 - 6 | 777 | 125 | ||
XM_012101611.3 | XP_011957001.1 | 4 - 5 | 722 | 125 | ||
XM_012101627.3 | XP_011957017.1 | 4 - 5 | 580 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |