Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114118084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978901.1 | XP_027834702.1 | 4 - 5 | 521 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |