Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103608.2 | XP_014959094.2 | 3 - 4 | 414 | 68 | |
XM_027960788.1 | XP_027816589.1 | 3 - 4 | 414 | 68 |