Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972684.1 | XP_027828485.1 | 8 - 9 | 890 | 108 | |
XM_027972685.1 | XP_027828486.1 | 8 - 9 | 890 | 108 | ||
XM_027972686.1 | XP_027828487.1 | 8 - 9 | 890 | 108 | ||
XM_027972687.1 | XP_027828488.1 | 8 - 9 | 890 | 108 | ||
XM_027972688.1 | XP_027828489.1 | 8 - 9 | 890 | 108 | ||
XM_027972689.1 | XP_027828490.1 | 8 - 9 | 890 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |