Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006863.4 | XP_004006912.1 | 3 - 4 | 321 | 127 | |
XM_012175354.3 | XP_012030744.1 | 3 - 4 | 250 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Ovis aries |