Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190547.3 | XP_012045937.2 | 5 - 6 | 1524 | 64 | |
XM_027978471.1 | XP_027834272.1 | 5 - 6 | 1524 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |