Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956674.1 | XP_027812475.1 | 3 - 4 | 701 | 76 | |
XM_027956675.1 | XP_027812476.1 | 3 - 4 | 676 | 76 | ||
XM_027956676.1 | XP_027812477.1 | 3 - 4 | 701 | 76 | ||
XM_027956677.1 | XP_027812478.1 | 3 - 4 | 701 | 76 | ||
XM_027956678.1 | XP_027812479.1 | 3 - 4 | 701 | 76 |