Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964741.1 | XP_027820542.1 | 3 - 4 | 330 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |