Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972904.1 | XP_027828705.1 | 10 - 11 | 1200 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |