Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016413.3 | XP_004016462.1 | 6 - 7 | 720 | 67 | |
XM_012096242.3 | XP_011951632.1 | 6 - 7 | 744 | 67 | ||
XM_012096243.3 | XP_011951633.1 | 8 - 9 | 854 | 67 | ||
XM_027979635.1 | XP_027835436.1 | 7 - 8 | 796 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |