Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973894.1 | XP_027829695.1 | 2 - 3 | 683 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |