Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961528.1 | XP_027817329.1 | 3 | 774 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |