Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022128.4 | XP_004022177.1 | 4 - 5 | 360 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |