Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002056.4 | XP_004002105.2 | 1 - 2 | 176 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |