Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966316.1 | XP_027822117.1 | 7 - 8 | 887 | 108 | |
XM_027966317.1 | XP_027822118.1 | 7 - 8 | 890 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |