Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177923.3 | XP_012033313.2 | 4 - 5 | 532 | 82 | |
XM_027970662.1 | XP_027826463.1 | 5 - 6 | 607 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |