Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957162.1 | XP_027812963.1 | 5 - 6 | 3723 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |