Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012186587.3 | XP_012041977.1 | 4 - 5 | 620 | 81 | |
XM_027974026.1 | XP_027829827.1 | 4 - 5 | 620 | 81 | ||
XM_027974027.1 | XP_027829828.1 | 4 - 5 | 617 | 81 | ||
XM_027974028.1 | XP_027829829.1 | 4 - 5 | 620 | 81 | ||
XM_027974029.1 | XP_027829830.1 | 4 - 5 | 620 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |