Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965894.1 | XP_027821695.1 | 12 - 13 | 1061 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |