Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098906.2 | XP_014954392.2 | 5 - 6 | 484 | 67 | |
XM_027974287.1 | XP_027830088.1 | 5 - 6 | 692 | 67 | ||
XM_027974288.1 | XP_027830089.1 | 5 - 6 | 454 | 67 | ||
XM_027974289.1 | XP_027830090.1 | 4 - 5 | 407 | 67 |