Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959295.1 | XP_027815096.1 | 8 - 9 | 2582 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |