Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970350.1 | XP_027826151.1 | 4 - 5 | 3445 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |