Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101104313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177654.3 | XP_012033044.1 | 4 - 5 | 501 | 113 | |
XM_027969330.1 | XP_027825131.1 | 4 - 5 | 472 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |