Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021042.4 | XP_004021091.1 | 3 - 4 | 1034 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |