Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009653.4 | XP_004009702.2 | 8 - 9 | 1231 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |