Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968065.1 | XP_027823866.1 | 5 - 6 | 710 | 108 | |
XM_027968066.1 | XP_027823867.1 | 5 - 6 | 790 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |