Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017345378.1 | XP_017200867.1 | 3 - 4 | 569 | 88 | |
GenBank mRNA | AB217855.1 | - | 2 - 3 | 360 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Inventoried Probe spans exons |