Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718264.3 | XP_002718310.1 | 4 - 5 | 572 | 71 | |
GenBank mRNA | AY575213.1 | - | 1 - 2 | 44 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |