Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KERA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711384.3 | XP_002711430.1 | 3 - 4 | 1037 | 70 | |
GenBank mRNA | DQ239829.1 | - | 1 - 2 | 481 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |