Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195721.1 | NP_001182650.1 | 9 - 10 | 1560 | 74 | |
XM_008263299.2 | XP_008261521.1 | 10 - 11 | 2039 | 74 | ||
XM_008263300.2 | XP_008261522.1 | 10 - 11 | 1967 | 74 | ||
GenBank mRNA | U04301.1 | - | 9 - 10 | 998 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons |